missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitrotoluene, 98%, Thermo Scientific™

Product Code. 15720799 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15720799 25 g Glass bottle 25g
10001090 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15720799 Supplier Thermo Scientific Acros Supplier No. 119520250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-10-6
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.13
MDL Number MFCD00007200
InChI Key SKWTUNAAJNDEIK-UHFFFAOYSA-N
Synonym 4-fluoro-2-nitrotoluene, 2-nitro-4-fluorotoluene, benzene, 4-fluoro-1-methyl-2-nitro, toluene, 4-fluoro-2-nitro, 4-fluoro-2-nitrobenzene, 5-fluoro-2-methylnitrobenzene, 4-fluoro-2-nitro toluene, 4-fluoro-1-methyl-2-nitro-benzene, 4-fluoronitrotoluene, zlchem 482
PubChem CID 67965
IUPAC Name 4-fluoro-1-methyl-2-nitrobenzene
SMILES CC1=C(C=C(C=C1)F)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 27°C
Color Amber to Yellow
Boiling Point 138°C to 139°C (83.0 mmHg)
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H3(NO2)F
Refractive Index 1.5218 to 1.5238
Quantity 25 g
Beilstein 05, I, 161
Formula Weight 155.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Fluoro-2-nitrotoluene, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.