missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methylbenzoic acid, 97%, Thermo Scientific™

Product Code. 11461117
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461117 5 g 5g
11451117 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11461117 Supplier Thermo Scientific Alfa Aesar Supplier No. B24112.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403-15-6
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD01631424
InChI Key VOCCEVKUXUIHOI-UHFFFAOYSA-N
Synonym 4-fluoro-m-toluic acid, 4-fluoro-3-methylbenzoicacid, 3-methyl-4-fluorobenzoic acid, 4-fluoro-3-methyl-benzoic acid, 3-methyl-4-fluoro benzoic acid, benzoic acid, 4-fluoro-3-methyl, pubchem1397, acmc-1aii0, ksc494s2d, rarechem al bo 2279
PubChem CID 242828
IUPAC Name 4-fluoro-3-methylbenzoic acid
SMILES CC1=C(C=CC(=C1)C(=O)O)F

Specifications

Melting Point 166°C to 168°C
Quantity 5 g
Beilstein 1936531
Formula Weight 154.14
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-methylbenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.