missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methylphenol, Thermo Scientific™

Product Code. 10102814 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10102814 5 g Glass bottle 5g
10480704 1 g Glass bottle 1g
10577054 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10102814 Supplier Thermo Scientific Acros Supplier No. 311950050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 452-70-0
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00042296
InChI Key RVYGYYVGWSCWGY-UHFFFAOYSA-N
Synonym 4-fluoro-m-cresol, 3-methyl-4-fluorophenol, 2-fluoro-5-hydroxytoluene, 4-fluoro-3-methyl-phenol, phenol, 4-fluoro-3-methyl, 4-fluoro-3-methylphenol, pubchem1501, 3-methyl-4-fluoro phenol, 4-fluoro-3-methyl phenol, acmc-209k1i
PubChem CID 2737394
IUPAC Name 4-fluoro-3-methylphenol
SMILES CC1=C(C=CC(=C1)O)F

Specifications

Melting Point 32.0°C
Color Colorless to Yellow
Flash Point 97°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CH3C6H3(F)OH
Refractive Index 1.5125 to 1.5145 (20°C, 589nm)
Quantity 5 g
Formula Weight 126.13
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 4-Fluoro-3-methylphenol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluoro-3-methylphenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.