missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitrobenzenesulfonyl chloride, TRC

Product Code. 30390495
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390495 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390495 Supplier Toronto Research Chemicals Supplier No. TRCF5976001G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6668-56-0
Molecular Formula C6H3ClFNO4S
Quantity 1 g
Synonym 4-Fluoro-3-nitrobenzene-1-sulfonyl chloride
InChI Formula InChI=1S/C6H3ClFNO4S/c7-14(12,13)4-1-2-5(8)6(3-4)9(10)11/h1-3H
SMILES [O-][N+](=O)c1cc(ccc1F)S(=O)(=O)Cl
Molecular Weight (g/mol) 239.61
Chemical Name or Material 4-Fluoro-3-nitrobenzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.