missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-7-nitro-2,1,3-benzoxadiazole, TRC

Product Code. 30134554
Change view
Click to view available options
Quantity:
5 mg
25 mg
250 mg
Unit Size:
250mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134554 25 mg 25mg
30136296 250 mg 250mg
30007550 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134554 Supplier Toronto Research Chemicals Supplier No. TRCF59470025MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29270-56-2
Molecular Formula C6 H2 F N3 O3
Molecular Weight (g/mol) 183.1
IUPAC Name 4-fluoro-7-nitro-2,1,3-benzoxadiazole
SMILES [O-][N+](=O)c1ccc(F)c2nonc12

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C6H2FN3O3/c7-3-1-2-4(10(11)12)6-5(3)8-13-9-6/h1-2H
Formula Weight 183.008
Chemical Name or Material 4-Fluoro-7-nitro-2,1,3-benzoxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.