missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorobenzaldehyde-2,3,5,6-d4, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 4-Fluorobenzaldehyde-2,3,5,6-d4 |
| CAS | 93111-27-4 |
| Molecular Formula | C7 D4 H F O |
| Quantity | 0.25 g |
| Synonym | p-Fluorobenzaldehyde-d4, p-Fluorobenzenecarboxaldehyde-d4; |
| InChI Formula | InChI=1 S/C7H5FO/c8-7-3-1-6(5-9)2-4-7/h1-5 H/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(C=O)c([2 H])c([2 H])c1F |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-fluorobenzaldehyde |
| Molecular Weight (g/mol) | 128.14 |
| Formula Weight | 128.0575 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction