missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoronitrobenzene-d4, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 4-Fluoronitrobenzene-D4 |
| CAS | 1219802-01-3 |
| Molecular Formula | C6 D4 F N O2 |
| Quantity | 0.25 g |
| Synonym | Benzene-1,2,4,5-d4, 3-fluoro-6-nitro- (ACI), 3-Fluoro-6-nitrobenzene-1,2,4,5-d4 (ACI), 4-Fluoronitrobenzene-d4, 4-Fluoronitrobenzene-D4, 4-Nitrofluorobenzene-d4 |
| InChI Formula | InChI=1 S/C6H4FNO2/c7-5-1-3-6(4-2-5)8(9)10/h1-4 H/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1F)[N+](=O)[O-] |
| IUPAC Name | 1,2,4,5-tetradeuterio-3-fluoro-6-nitrobenzene |
| Molecular Weight (g/mol) | 145.1245 |
| Formula Weight | 145.0477 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction