Learn More
4-Fluorophenyl methyl sulfone, 98%
Product Code. 11485283
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485283 5 g 5g
11495283 25 g 25g
11405293 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11485283 Supplier Thermo Scientific Alfa Aesar Supplier No. A12863.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
CAS: 455-15-2 | C7H7FO2S | 174.189 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 455-15-2
Molecular Formula C7H7FO2S
Molecular Weight (g/mol) 174.189
MDL Number MFCD00039753
InChI Key DPJHZJGAGIWXTD-UHFFFAOYSA-N
Synonym 4-fluorophenyl methyl sulfone, 1-fluoro-4-methylsulfonyl benzene, benzene, 1-fluoro-4-methylsulfonyl, 1-fluoro-4-methylsulphonyl benzene, 1-fluoro-4-methanesulfonylbenzene, 4-fluorophenyl methyl sulphone, 4-fluorophenylmethylsulfone, sulfone, p-fluorophenyl methyl, methyl 4-fluorophenyl sulfone, 1-fluoro-4-methylsulfonyl-benzene
PubChem CID 67994
IUPAC Name 1-fluoro-4-methylsulfonylbenzene
SMILES CS(=O)(=O)C1=CC=C(C=C1)F
Specifications
Melting Point 76°C to 81°C
Quantity 5 g
Beilstein 2045674
Formula Weight 174.19
Percent Purity 98%
Chemical Name or Material 4-Fluorophenyl methyl sulfone