missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenyl methyl sulfone, 99%, Thermo Scientific™

Product Code. 10717593 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10717593 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10717593 Supplier Thermo Scientific Acros Supplier No. 303360010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 455-15-2
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C7H7FO2S
Linear Formula FC6H4SO2CH3
MDL Number MFCD00039753
Quantity 1g
Synonym 4-fluorophenyl methyl sulfone, 1-fluoro-4-methylsulfonyl benzene, benzene, 1-fluoro-4-methylsulfonyl, 1-fluoro-4-methylsulphonyl benzene, 1-fluoro-4-methanesulfonylbenzene, 4-fluorophenyl methyl sulphone, 4-fluorophenylmethylsulfone, sulfone, p-fluorophenyl methyl, methyl 4-fluorophenyl sulfone, 1-fluoro-4-methylsulfonyl-benzene
InChI Key DPJHZJGAGIWXTD-UHFFFAOYSA-N
SMILES CS(=O)(=O)C1=CC=C(C=C1)F
IUPAC Name 1-fluoro-4-methylsulfonylbenzene
Molecular Weight (g/mol) 174.189
PubChem CID 67994
Formula Weight 174.19
Percent Purity 99%
Chemical Name or Material 4-Fluorophenyl methyl sulfone, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.