missing translation for 'onlineSavingsMsg'
Learn More

4-Hexylbenzoic acid, 96%, Thermo Scientific™

Product Code. 10491851 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10491851 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10491851 Supplier Thermo Scientific Acros Supplier No. 258880025

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 21643-38-9
CAS Min % 95.0
CAS Max % 100.0
Melting Point 94°C to 99°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Molecular Formula C13H18O2
Linear Formula CH3(CH2)4CH2C6H4COOH
MDL Number MFCD00013997
Quantity 2.5 g
Synonym benzoic acid, 4-hexyl, 4-n-hexylbenzoic acid, p-hexylbenzoic acid, benzoic acid, p-hexyl, p-n-hexylbenzoic acid, 4n-hexylbenzoic acid, maybridge1_002028, para-n-hexyl benzoic acid, acmc-1chu5, ksc201s3p
InChI Key CPEPWESLFZVUEP-UHFFFAOYSA-N
SMILES CCCCCCC1=CC=C(C=C1)C(=O)O
IUPAC Name 4-hexylbenzoic acid
Molecular Weight (g/mol) 206.285
PubChem CID 30725
Formula Weight 206.28
Percent Purity ≥95%
Physical Form Slightly Crystalline Powder or Flakes
Chemical Name or Material 4-Hexylbenzoic acid, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Hexylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap and water.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.