missing translation for 'onlineSavingsMsg'
Learn More

4’-Hydroxy Atomoxetine, TRC

Product Code. 17964875
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17964875 1 mg 1mg
17954245 10 mg 10mg
17933235 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17964875 Supplier Toronto Research Chemicals Supplier No. TRCH8025001MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 435293-66-6
Molecular Formula C17 H21 N O2
Molecular Weight (g/mol) 271.35
Synonym Phenol, 3-methyl-4-[(1R)-3-(methylamino)-1-phenylpropoxy]-, 3-Methyl-4-[(1R)-3-(methylamino)-1-phenylpropoxy]phenol, 4-Hydroxyatomoxetine
IUPAC Name 3-methyl-4-[(1R)-3-(methylamino)-1-phenylpropoxy]phenol
SMILES CNCC[C@@H](Oc1ccc(O)cc1C)c2ccccc2

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C17H21NO2/c1-13-12-15(19)8-9-16(13)20-17(10-11-18-2)14-6-4-3-5-7-14/h3-9,12,17-19H,10-11H2,1-2H3/t17-/m1/s1
Formula Weight 271.1572
Percent Purity >95%
Chemical Name or Material 4-Hydroxy Atomoxetine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.