missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-N-isopropyl-N-methyltryptamine, TRC

Product Code. 30403985
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30403985 25 mg -
30395226 10 mg -
30401567 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403985 Supplier Toronto Research Chemicals Supplier No. TRCH94288025MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77872-43-6
Molecular Formula C14 H20 N2 O
Molecular Weight (g/mol) 232.32
Synonym 1H-Indol-4-ol, 3-[2-[methyl(1-methylethyl)amino]ethyl]-, 3-[2-[Methyl(1-methylethyl)amino]ethyl]-1H-indol-4-ol, 4-Hydroxy-N-isopropyl-N-methyltryptamine, 4-HO-MiPT, 4-Hydroxy-N-methyl-N-isopropyltryptamine
IUPAC Name 3-[2-[methyl(propan-2-yl)amino]ethyl]-1H-indol-4-ol
SMILES CC(C)N(C)CCc1c[nH]c2cccc(O)c12

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C14H20N2O/c1-10(2)16(3)8-7-11-9-15-12-5-4-6-13(17)14(11)12/h4-6,9-10,15,17H,7-8H2,1-3H3
Formula Weight 232.1576
Chemical Name or Material 4-hydroxy MiPT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.