missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-tetramisole, TRC

Product Code. 17900845
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
Unit Size:
1mg
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17900845 1 mg 1mg
17902575 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17900845 Supplier Toronto Research Chemicals Supplier No. TRCH5952701MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6649-26-9
Molecular Formula C11 H12 N2 O S
Molecular Weight (g/mol) 220.29
Synonym Phenol, 4-(2,3,5,6-tetrahydroimidazo[2,1-b]thiazol-6-yl)-, Phenol, p-(2,3,5,6-tetrahydroimidazo[2,1-b]thiazol-6-yl)- (7CI,8CI), 4-(2,3,5,6-Tetrahydroimidazo[2,1-b]thiazol-6-yl)phenol, Imidazo[2,1-b]thiazole, phenol deriv.
IUPAC Name 4-(2,3,5,6-tetrahydroimidazo[2,1-b][1,3]thiazol-6-yl)phenol
SMILES Oc1ccc(cc1)C2CN3CCSC3=N2

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C11H12N2OS/c14-9-3-1-8(2-4-9)10-7-13-5-6-15-11(13)12-10/h1-4,10,14H,5-7H2
Formula Weight 220.067
Chemical Name or Material Tetramisole-4-hydroxy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.