missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzenesulfonic Acid (85%), TRC

Product Code. 30407252
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30407252 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30407252 Supplier Toronto Research Chemicals Supplier No. TRCH80780510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Hydroxybenzenesulfonic Acid
CAS 98-67-9
Molecular Formula C6H6O4S
Quantity 10 g
Synonym 4-Hydroxyphenylsulfonic acid, 4-Phenolsulfonic acid, NSC 227908, p-Hydroxybenzenesulfonic acid, p-Phenolsulfonic acid, p-Sulfophenol
InChI Formula InChI=1S/C6H6O4S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4,7H,(H,8,9,10)
SMILES CCC(C(N)=O)(C(N)=O)c1ccccc1
IUPAC Name 4-hydroxybenzenesulfonic acid
Molecular Weight (g/mol) 174.17
Formula Weight 173.9987
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.