missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxychrysene, TRC

Product Code. 30136106
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136106 2.5 mg 2.5mg
30138531 10 mg 10mg
30010382 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136106 Supplier Toronto Research Chemicals Supplier No. TRCH9053452.5MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63019-40-9
Molecular Formula C18H12O
Molecular Weight (g/mol) 244.29
Synonym 4-Chrysenol
IUPAC Name chrysen-4-ol
SMILES Oc1cccc2ccc3c4ccccc4ccc3c12

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C18H12O/c19-17-7-3-5-13-9-10-15-14-6-2-1-4-12(14)8-11-16(15)18(13)17/h1-11,19H
Formula Weight 244.0888
Chemical Name or Material 4-Hydroxychrysene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.