missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)phenylboronic acid, 97%, Thermo Scientific™

Product Code. 10364642 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10364642 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10364642 Supplier Thermo Scientific Acros Supplier No. 369470010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 59016-93-2
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C7H9BO3
MDL Number MFCD00792672
Quantity 1g
Synonym 4-hydroxymethyl phenylboronic acid, 4-hydroxymethyl benzeneboronic acid, 4-hydroxymethyl phenyl boronic acid, 4-hydroxymethylphenyl boronic acid, 4-hydroxymethylphenylboronic acid, 4-boronobenzyl alcohol, p-hydroxymethyl phenylboronic acid, 4-hydroxymethyl phenyl boranediol
InChI Key PZRPBPMLSSNFOM-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)CO)(O)O
IUPAC Name [4-(hydroxymethyl)phenyl]boronic acid
Molecular Weight (g/mol) 151.956
PubChem CID 2734706
Formula Weight 151.96
Percent Purity ≥96.0%
Chemical Name or Material 4-(Hydroxymethyl)phenylboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.