missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenyl 4-Isopropoxyphenyl Sulfone, TRC

Product Code. 30137037
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137037 25 g 25g
30135408 100 g 100g
30005963 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137037 Supplier Toronto Research Chemicals Supplier No. TRCH94957025G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95235-30-6
Molecular Formula C15 H16 O4 S
Molecular Weight (g/mol) 292.35
Synonym 4-[[4-(1-Methylethoxy)phenyl]sulfonyl]phenol, 4-Hydroxy-4'-isopropoxy phenyl sulfone, 4-Hydroxy-4'-isopropoxydiphenyl sulfone, 4-Hydroxyphenyl 4-isopropoxyphenyl sulfone, 4-Isopropoxy-4'-hydroxydiphenyl sulfone, D 8, D 8 (sulfone), F 647, NYDS
IUPAC Name 4-(4-propan-2-yloxyphenyl)sulfonylphenol
SMILES CC(C)Oc1ccc(cc1)S(=O)(=O)c2ccc(O)cc2

Specifications

Quantity 25 g
InChI Formula InChI=1S/C15H16O4S/c1-11(2)19-13-5-9-15(10-6-13)20(17,18)14-7-3-12(16)4-8-14/h3-11,16H,1-2H3
Formula Weight 292.0769
Percent Purity >95%
Chemical Name or Material Bisphenol S-monoisopropyl ether
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.