missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzyl alcohol, 98%, ACROS Organics™

Product Code. 11954651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11954651 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11954651 Supplier Thermo Scientific Acros Supplier No. 394480050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 18282-51-4
CAS Min % 97.5
CAS Max % 100.0
Melting Point 72.0°C to 75.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H7IO
Linear Formula IC6H4CH2OH
Quantity 5 g
Synonym 4-iodobenzyl alcohol, 4-iodophenyl methanol, p-iodobenzyl alcohol, benzenemethanol, 4-iodo, benzyl alcohol, p-iodo, 4-iodobenzylalcohol, p-iodobenzylalcohol, 4-iodobenzyl-alcohol, 4-iodobenzyl zlcohol, p-lodo-benzyl alcohol
InChI Key CNQRHSZYVFYOIE-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CO)I
IUPAC Name (4-iodophenyl)methanol
Molecular Weight (g/mol) 234.036
PubChem CID 29012
Formula Weight 234.03
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodobenzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 4-Iodobenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.