missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzylamine, 97%

Product Code. 11388416
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388416 1 g 1g
11398416 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388416 Supplier Thermo Scientific Alfa Aesar Supplier No. L17450.03
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 39959-59-6 | C7H8IN | 233.052 g/mol

4-Iodobenzylamine is used to prepare 2-bromo-N-(4-iodo-benzyl)-acetamide in the presence of bromoacetyl bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzylamine is used to prepare 2-bromo-N-(4-iodo-benzyl)-acetamide in the presence of bromoacetyl bromide.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to air and light. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39959-59-6
Molecular Formula C7H8IN
Molecular Weight (g/mol) 233.052
MDL Number MFCD00047933
InChI Key KCGZGJOBKAXVSU-UHFFFAOYSA-N
Synonym 4-iodophenyl methanamine, 4-iodobenzylamine, 4-iodo-benzylamine, 4-iodobenzyl amine, benzenemethanamine, 4-iodo, 1-4-iodophenyl methanamine, 4-iodophenyl methylamine, 4-iodobenzylamine, hydrochloride salt, acmc-20a5sy, p-iodobenzylamine
PubChem CID 2737533
IUPAC Name (4-iodophenyl)methanamine
SMILES C1=CC(=CC=C1CN)I

Specifications

Melting Point 46°C to 48°C
Boiling Point 113°C to 115°C (4 mmHg)
Quantity 1 g
UN Number UN3259
Beilstein 2689605
Sensitivity Air and light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 233.05
Percent Purity 97%
Chemical Name or Material 4-Iodobenzylamine
compliance-icons
Product Identifier
  • 4-Iodobenzylamine
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.