missing translation for 'onlineSavingsMsg'
Learn More

4-Isobutylbenzeneboronic acid, 98%

Product Code. 15429367
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15429367 5 g 5g
15419367 1 g 1g
15439367 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15429367 Supplier Thermo Scientific Alfa Aesar Supplier No. H52603.06
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 153624-38-5 | C10H15BO2 | 178.038 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153624-38-5
Molecular Formula C10H15BO2
Molecular Weight (g/mol) 178.038
MDL Number MFCD01632198
InChI Key YZSPHVWALUJQNK-UHFFFAOYSA-N
Synonym 4-isobutylphenylboronic acid, 4-isobutylbenzeneboronic acid, 4-isobutylphenyl boronic acid, 4-2-methylpropyl phenylboronic acid, 4-2-methylpropyl phenyl boronic acid, 4-2-methylpropyl phenyl boranediol, boronic acid, 4-2-methylpropyl phenyl, boronic acid,b-4-2-methylpropyl phenyl, 4-iso-butylphenylboronic acid, pubchem20832
PubChem CID 2773472
IUPAC Name [4-(2-methylpropyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)CC(C)C)(O)O

Specifications

Melting Point 117°C to 120°C
Quantity 5 g
Formula Weight 178.04
Percent Purity 98%
Chemical Name or Material 4-Isobutylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.