missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropoxybenzoic acid, 99%

Product Code. 11446917
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446917 10 g 10g
11456917 50 g 50g
11466917 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11446917 Supplier Thermo Scientific Alfa Aesar Supplier No. A14351.09
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 13205-46-4 | C10H12O3 | 180.203 g/mol

4-Isopropoxybenzoic acid is used to prepare 5-(4-isopropoxyphenyl)-N-phenyl-1,3,4-thiadiazol-2-amine by reacting with N-phenylhydrazinecarbothioamide in the presence of phosphorusoxychloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Isopropoxybenzoic acid is used to prepare 5-(4-isopropoxyphenyl)-N-phenyl-1,3,4-thiadiazol-2-amine by reacting with N-phenylhydrazinecarbothioamide in the presence of phosphorusoxychloride.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13205-46-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00044318
InChI Key ZVERWTXKKWSSHH-UHFFFAOYSA-N
PubChem CID 72972
IUPAC Name 4-propan-2-yloxybenzoic acid
SMILES CC(C)OC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 163°C to 167°C
Odor Odorless
Quantity 10 g
Synonym 4-isopropoxybenzoic acid, benzoic acid, 4-1-methylethoxy, 4-propan-2-yloxy benzoic acid, 4-isopropoxybenzoicacid, 4-isopropoxy-benzoic acid, 4-iso-propyloxybenzoic acid, rarechem al be 0294, 4-isopropyloxybenzoic acid, 4-1-methylethoxy benzoic acid, benzoic acid,4-1-methylethoxy
Solubility Information Insoluble in water.
Formula Weight 180.2
Percent Purity 99%
Chemical Name or Material 4-Isopropoxybenzoic acid
compliance-icons
Product Identifier
  • 4-Isopropoxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 4-Isopropoxybenzoicacid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.