missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylphenyl Diphenyl Phosphate, TRC

Product Code. 30133572
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133572 1 g 1g
30012819 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133572 Supplier Toronto Research Chemicals Supplier No. TRCI8729451G
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55864-04-5
Molecular Formula C21H21O4P
Molecular Weight (g/mol) 368.36
Synonym p-Cumenyl Phenyl Phosphate, Diphenyl 4-isopropylphenyl Phosphate, p-Isopropylphenyl Diphenyl Phosphate
IUPAC Name diphenyl (4-propan-2-ylphenyl) phosphate
SMILES O=P(OC1=CC=CC=C1)(OC2=CC=CC=C2)OC3=CC=C(C(C)C)C=C3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C21H21O4P/c1-17(2)18-13-15-21(16-14-18)25-26(22,23-19-9-5-3-6-10-19)24-20-11-7-4-8-12-20/h3-17H,1-2H3
Formula Weight 368.1177
Chemical Name or Material 4-Isopropylphenyl Diphenyl Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.