Learn More
4-Maleimidobutyric Acid N-Succinimidyl Ester, TRC
Specifications
Specifications
| CAS | 80307-12-6 |
| Molecular Formula | C12 H12 N2 O6 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C12H12N2O6/c15-8-3-4-9(16)13(8)7-1-2-12(19)20-14-10(17)5-6-11(14)18/h3-4H,1-2,5-7H2 |
| SMILES | O=C(CCCN1C(=O)C=CC1=O)ON2C(=O)CCC2=O |
| IUPAC Name | (2,5-dioxopyrrolidin-1-yl) 4-(2,5-dioxopyrrol-1-yl)butanoate |
| Molecular Weight (g/mol) | 280.23 |
| Formula Weight | 280.0695 |
| Chemical Name or Material | 4-Maleimidobutyric Acid N-Succinimidyl Ester |