missing translation for 'onlineSavingsMsg'
Learn More

4-Mercaptopyridine, TRC

Product Code. 30476077
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30476077 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30476077

Brand: Toronto Research Chemicals TRCM2255001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Mercaptopyridine
CAS 4556-23-4
Molecular Formula C5H5NS
Quantity 1 g
Synonym 4-Pyridinethiol, 4-Thiopyridine, 4-Sulfanylpyridine, NSC 76036
InChI Formula InChI=1S/C5H5NS/c7-5-1-3-6-4-2-5/h1-4H,(H,6,7)
SMILES Sc1ccncc1
IUPAC Name pyridine-4-thiol
Molecular Weight (g/mol) 111.16
Formula Weight 111.0143
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.