Learn More
4-Methoxy-2,3,6-trimethylbenzaldehyde, TRC
Product Code. 30480099
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30480099 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30480099 Supplier Toronto Research Chemicals Supplier No. TRCM2697001G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Methoxy-2,3,6-trimethylbenzaldehyde
CAS 54344-92-2
Molecular Formula C11 H14 O2
Quantity 1 g
Synonym Benzaldehyde','4-methoxy-2,3,6-trimethyl-, p-Anisaldehyde','2,3,6-trimethyl- (7CI), 4-Methoxy-2,3,6-trimethylbenzaldehyde, 2,3,6-Trimethyl-p-anisaldehyde
InChI Formula InChI=1S/C11H14O2/c1-7-5-11(13-4)9(3)8(2)10(7)6-12/h5-6H,1-4H3
SMILES COc1cc(C)c(C=O)c(C)c1C
IUPAC Name 4-methoxy-2,3,6-trimethylbenzaldehyde
Molecular Weight (g/mol) 178.23
Formula Weight 178.0994