missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyaniline-2,3,5,6-d4, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.10g
0.25g
Specifications
Specifications
| Chemical Name or Material | 2,3,5,6-Tetradeuterio-4-methoxyaniline |
| CAS | 1219798-55-6 |
| Molecular Formula | C7 D4 H5 N O |
| Quantity | 0.1 g |
| Synonym | Benzen-2,3,5,6-d4-amine, 4-methoxy- (ACI), 4-Methoxybenzen-2,3,5,6-d4-amine (ACI), 2,3,5,6-Tetradeuterio-4-methoxyaniline, 4-Methoxyaniline-2,3,5,6-d4, 4-Methoxyaniline-2,3,5,6-D4, p-Anisidine-d4 |
| InChI Formula | InChI=1 S/C7H9NO/c1-9-7-4-2-6(8)3-5-7/h2-5 H,8H2,1H3/i2D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(OC)c([2 H])c([2 H])c1N |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-methoxyaniline |
| Molecular Weight (g/mol) | 127.177 |
| Formula Weight | 127.094 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction