missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzyl Chloride (Stabilized), TRC

Product Code. 30478465
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478465 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478465 Supplier Toronto Research Chemicals Supplier No. TRCM26110525G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methoxybenzyl Chloride
CAS 824-94-2
Molecular Formula C8 H9 Cl O
Quantity 25 g
Synonym p-(Chloromethyl)-anisole, 1-(Chloromethyl)-4-(methyloxy)benzene, 1-(Chloromethyl)-4-methoxybenzene, 4-(Chloromethyl)-1-methoxybenzene, 4-(Chloromethyl)anisole, 4-(Chloromethyl)phenyl Methyl Ether, 4-Methoxybenzyl Chloride, 4-Methyloxybenzyl Chloride, NSC 172955, p-(Chloromethyl)(methoxy)benzene, p-(Chloromethyl)anisole, p-Anisyl Chloride, p-Methoxybenzyl Chloride, α-Chloro-4-methoxytoluene, α-Chloro-p-methoxytoluene, PMB-Cl
InChI Formula InChI=1S/C8H9ClO/c1-10-8-4-2-7(6-9)3-5-8/h2-5H,6H2,1H3
SMILES COc1ccc(CCl)cc1
IUPAC Name 1-(chloromethyl)-4-methoxybenzene
Molecular Weight (g/mol) 156.61
Formula Weight 156.0342
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.