missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyestrone-1,2,16,16-d4, CDN
Click to view available options
Quantity:
0.005 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 4-Methoxyestrone-1,2,16,16-D4 |
| Molecular Formula | C19 D4 H20 O3 |
| Quantity | 0.005 g |
| Synonym | 4-Methoxyestrone-1,2,16,16-d4, 3-Hydroxy-4-methoxyestra-1,3,5(10)-trien-17-one-1,2,16,16-d4 |
| InChI Formula | InChI=1 S/C19H24O3/c1-19-10-9-12-11-5-7-16(20)18(22-2)14(11)4-3-13(12)15(19)6-8-17(19)21/h5,7,12-13,15,20 H,3-4,6,8-10H2,1-2H3/t12-,13-,15+,19+/m1/s1/i5D,7 D,8D2 |
| SMILES | [2 H]c1c([2 H])c2[C@H]3 CC[C@@]4(C)[C@@H](CC([2 H])([2 H])C4=O)[C@@H]3 CCc2c(OC)c1O |
| IUPAC Name | (8 R,9 S,13 S,14 S)-1,2,16,16-tetradeuterio-3-hydroxy-4-methoxy-13-methyl-6,7,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthren-17-one |
| Molecular Weight (g/mol) | 304.4167 |
| Formula Weight | 304.1977 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction