Learn More
4-Methoxyphenylacetonitrile, 98%
Product Code. 11417337
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417337 25 g 25g
11427337 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11417337 Supplier Thermo Scientific Alfa Aesar Supplier No. A15056.14
 Restricted Item
In Stock
Add to basket
CAS: 104-47-2 | C9H9NO | 147.177 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 104-47-2
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00001919
InChI Key PACGLQCRGWFBJH-UHFFFAOYSA-N
Synonym 4-methoxyphenylacetonitrile, 2-4-methoxyphenyl acetonitrile, 4-methoxybenzyl cyanide, 4-methoxyphenyl acetonitrile, p-methoxyphenylacetonitrile, benzeneacetonitrile, 4-methoxy, p-methoxybenzyl cyanide, anisylacetonitrile, 4-anisyl cyanide, p-methoxybenzeneacetonitrile
PubChem CID 66031
IUPAC Name 2-(4-methoxyphenyl)acetonitrile
SMILES COC1=CC=C(C=C1)CC#N
Specifications
Melting Point 8°C
Density 1.085
Boiling Point 285°C to 287°C
Flash Point 117°C (242°F)
Refractive Index 1.53
Quantity 25 g
UN Number UN3276
Beilstein 509162
Formula Weight 147.18
Percent Purity 98%
Chemical Name or Material 4-Methoxyphenylacetonitrile