missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylacetonitrile, TRC

Product Code. 30477201
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477201 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477201 Supplier Toronto Research Chemicals Supplier No. TRCM266380100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (4-Methoxyphenyl)acetonitrile
CAS 104-47-2
Molecular Formula C9 H9 N O
Quantity 100 g
Synonym Benzeneacetonitrile','4-methoxy-, Acetonitrile','(p-methoxyphenyl)- (6CI,7CI,8CI), 4-Methoxybenzeneacetonitrile, (4-Methoxyphenyl)acetonitrile, (p-Methoxyphenyl)acetonitrile, 1-(Cyanomethyl)-4-methoxybenzene, 2-(4-Methoxyphenyl)acetonitrile, 4-Methoxybenzyl cyanide, NSC 96, PAI 106, p-Anisylacetonitrile, p-Methoxybenzeneacetonitrile, p-Methoxybenzyl cyanide, p-Methoxybenzylnitrile
InChI Formula InChI=1S/C9H9NO/c1-11-9-4-2-8(3-5-9)6-7-10/h2-5H,6H2,1H3
SMILES COc1ccc(CC#N)cc1
IUPAC Name 2-(4-methoxyphenyl)acetonitrile
Molecular Weight (g/mol) 147.1739
Formula Weight 147.0684
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.