missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-tetralone, 97%, Thermo Scientific™

Product Code. 11495077
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495077 1 g 1g
11405087 5 g 5g
11415087 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495077 Supplier Thermo Scientific Alfa Aesar Supplier No. B21967.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Methyl-1-tetralone is an important intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19832-98-5
Molecular Formula C11H12O
Molecular Weight (g/mol) 160.22
MDL Number MFCD00001691
InChI Key SRLHDEROUKFEMJ-UHFFFAOYNA-N
Synonym 4-methyl-1-tetralone, 4-methyl-3,4-dihydronaphthalen-1 2h-one, 4-methyl-.alpha.-tetralone, 1 2h-naphthalenone, 3,4-dihydro-4-methyl, 4-methyl-1,2,3,4-tetrahydronaphthalen-1-one, 3,4-dihydro-4-methyl-1 2h-naphthalenone, 4-methyl-2,3,4-trihydronaphthalen-1-one, zlchem 108, pubchem11038, 4-methyltetralin-1-one
PubChem CID 89232
IUPAC Name 4-methyl-3,4-dihydro-2H-naphthalen-1-one
SMILES CC1CCC(=O)C2=CC=CC=C12

Specifications

Density 1.078
Boiling Point 95°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.56
Quantity 1 g
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 160.22
Percent Purity 97%
Chemical Name or Material 4-Methyl-1-tetralone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.