missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-(4-trifluoromethylphenyl)thiazole-5-carboxylic Acid Ethyl Ester, TRC

Product Code. 30466678
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30466678 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30466678 Supplier Toronto Research Chemicals Supplier No. TRCM330955100MG
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methyl-2-(4-trifluoromethylphenyl)thiazole-5-carboxylic Acid Ethyl Ester
CAS 175277-03-9
Molecular Formula C14H12F3NO2S
Quantity 100 mg
Synonym Ethyl 4-Methyl-2-[4-(trifluoromethyl)phenyl]thiazole-5-carboxylate
InChI Formula InChI=1S/C14H12F3NO2S/c1-3-20-13(19)11-8(2)18-12(21-11)9-4-6-10(7-5-9)14(15,16)17/h4-7H,3H2,1-2H3
SMILES CCOC(=O)c1sc(nc1C)c2ccc(cc2)C(F)(F)F
IUPAC Name ethyl 4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol) 315.31
Formula Weight 315.0541
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.