missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanone, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. p-7220840 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10087460 1 L 1 L
10484801 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10087460 Supplier JT Baker Supplier No. 8049.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Exceeds ACS Specifications

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00008938
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
Synonym Methyl Isobutyl Ketone , Isobutyl Methyl Ketone , Methyl iso-Butyl Ketone
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(C)=O

Specifications

Melting Point -84°C
Color Max. 15 (APHA)
Appearance Clear
Density 0.801 g/cm³ (20°C)
Quantity 1 L
Flash Point 16°C
Packaging Glass Bottle
Residue after Evaporation max. 0.002%
Assay Percent Range min. 99%
Water max. 0.1%
Boiling Point 117.4°C (1013 hPa)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.