missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pyrid-4-yl-1,3-thiazole-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10577654
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10577654 250 mg Amber Glass Bottle 250mg
10587654 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10577654 Supplier Thermo Scientific Maybridge Supplier No. CC58604CB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 892502-19-1
Molecular Formula C10H8N2OS
Molecular Weight (g/mol) 204.247
MDL Number MFCD09817496
InChI Key DBJGBXWDMYPPLF-UHFFFAOYSA-N
Synonym 4-methyl-2-pyridin-4-yl thiazole-5-carbaldehyde, 4-methyl-2-pyridin-4-yl-1,3-thiazole-5-carbaldehyde, 4-methyl-2-pyrid-4-yl-1,3-thiazole-5-carbaldehyde, 4-methyl-2-pyridin-4-yl-1,3-thiazole-5-carboxaldehyde, 5-formyl-4-methyl-2-pyridin-4-yl-1,3-thiazole, 5-thiazolecarboxaldehyde,4-methyl-2-4-pyridinyl, 4-methyl-2-4-pyridyl-1,3-thiazole-5-carbaldehyde
PubChem CID 24229586
IUPAC Name 4-methyl-2-pyridin-4-yl-1,3-thiazole-5-carbaldehyde
SMILES CC1=C(SC(=N1)C2=CC=NC=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 204.25
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-pyrid-4-yl-1, 3-thiazole-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.