missing translation for 'onlineSavingsMsg'
Learn More

4'-Methyl-3,4-dihydroxybenzophenone, Mikromol™

Product Code. 30050101
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050101 100 mg 100mg
30050030 25 mg 25mg
30457452 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30050101 Supplier Mikromol Supplier No. MM3329.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4'-Methyl-3,4-dihydroxybenzophenone
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4'-Methyl-3,4-dihydroxybenzophenone
CAS 1391053-03-4
Analyte or Component Names 4'-Methyl-3,4-dihydroxybenzophenone
Shipping Condition Room Temperature
Molecular Formula C14 H12 O3
Certifications/Compliance ISO 9001
Synonym 4'-Methyl-3,4-dihydroxybenzophenone, Tolcapone Related Compound A
InChI Formula InChI=1S/C14H12O3/c1-9-2-4-10(5-3-9)14(17)11-6-7-12(15)13(16)8-11/h2-8,15-16H,1H3
SMILES Cc1ccc(cc1)C(=O)c2ccc(O)c(O)c2
IUPAC Name (3,4-dihydroxyphenyl)-(4-methylphenyl)methanone
Molecular Weight (g/mol) 228.24
Quantity 100 mg
Formula Weight 228.0786 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.