missing translation for 'onlineSavingsMsg'
Learn More

4'-Methyl-3,4-dihydroxybenzophenone, Mikromol™

Product Code. 30457452
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457452 4 x 25 mg 100mg
30050101 100 mg 100mg
30050030 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457452 Supplier Mikromol Supplier No. MM3329.0200254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4'-Methyl-3,4-dihydroxybenzophenone, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4'-Methyl-3,4-dihydroxybenzophenone
CAS 1391053-03-4
Analyte or Component Names 4'-Methyl-3,4-dihydroxybenzophenone
Shipping Condition Room Temperature
Molecular Formula C14 H12 O3
Certifications/Compliance ISO 9001
Synonym 4'-Methyl-3,4-dihydroxybenzophenone, Tolcapone Related Compound A
InChI Formula InChI=1S/C14H12O3/c1-9-2-4-10(5-3-9)14(17)11-6-7-12(15)13(16)8-11/h2-8,15-16H,1H3
Solution Type Neat
SMILES Cc1ccc(cc1)C(=O)c2ccc(O)c(O)c2
IUPAC Name (3,4-dihydroxyphenyl)-(4-methylphenyl)methanone
Molecular Weight (g/mol) 228.24
Quantity 4 x 25 mg
Formula Weight 228.0786 g/mol
Show More Show Less
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.