Learn More
4-Methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific™
Product Code. 10289232
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10289232 250 mg Amber Glass Bottle 250mg
10548053 1 g Amber Glass Bottle 1g
11346825 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10289232 Supplier Thermo Scientific Maybridge Supplier No. CC13539CB
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Chemical Identifiers
CAS 519054-54-7
Molecular Formula C15H22BNO3
Molecular Weight (g/mol) 275.16
MDL Number MFCD04115377
InChI Key QRAOZQGIUIDZQZ-UHFFFAOYSA-N
Synonym 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine, 4-methyl-3,4-dihydro-2h-1,4-benzoxazin-7-ylboronic acid pinacol ester, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1,4-benzoxazine, 4-methyl-7-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-benzo 1,4 oxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2,-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-d ioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine
PubChem CID 2795301
IUPAC Name 4-methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1,4-benzoxazine
SMILES CN1CCOC2=C1C=CC(=C2)B1OC(C)(C)C(C)(C)O1
Specifications
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 275.15
Chemical Name or Material 4-Methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine