missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-d3-benzyl bromide, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 4-Methylbenzyl Bromide-D3 (4-methyl-D3) |
| CAS | 114563-29-0 |
| Molecular Formula | C8 D3 H6 Br |
| Quantity | 0.1 g |
| Synonym | Benzene, 1-(bromomethyl)-4-(methyl-d3)- (9 CI), 1-Bromomethyl-4-(methyl-d3)benzene, p-Methylbenzyl Bromide-d3, 1-(Methyl-d3)-4-bromomethylbenzene, 4-Methylbenzyl Bromide-α',α',α'-D3, α-Bromo-p-xylene-α',α',α'-d2, α-Bromo-p-xylene-α',α',α'-d2 |
| InChI Formula | InChI=1 S/C8H9Br/c1-7-2-4-8(6-9)5-3-7/h2-5 H,6H2,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])c1ccc(CBr)cc1 |
| IUPAC Name | 1-(bromomethyl)-4-(trideuteriomethyl)benzene |
| Molecular Weight (g/mol) | 188.0796 |
| Formula Weight | 187.0076 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction