missing translation for 'onlineSavingsMsg'
Learn More

4-Methylamino Phenol Sulphate, Extra Pure, SLR, Fisher Chemical

Product Code. 10446880 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic powder jar
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10446880 250 g Plastic powder jar 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10446880 Supplier Fisher Chemical Supplier No. M/5400/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 55-55-0
Melting Point 260°C
pH 3.5 to 4.5
Packaging Plastic powder jar
Molecular Formula C14H18N2O2·H2SO4
MDL Number 13140
Quantity 250 g
Formula Weight 344.38g/mol
Physical Form Solid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Methylaminophenol sulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.