missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzonitrile, Mikromol™

Product Code. 17913246
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17913246 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17913246 Supplier Mikromol Supplier No. MM1143.170100
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Methylbenzonitrile
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methylbenzonitrile (p-Tolunitrile)
CAS 104-85-8
Analyte or Component Names 4-Methylbenzonitrile (p-Tolunitrile)
Shipping Condition Room Temperature
Molecular Formula C8 H7 N
Certifications/Compliance ISO 9001
Synonym Benzonitrile, 4-methyl-, p-Tolunitrile (8CI), 4-Methylbenzonitrile, 1-Cyano-4-methylbenzene, 4-Cyanotoluene, 4-Methylcyanobenzene, 4-Methylphenyl cyanide, 4-Methylphenylnitrile, 4-Toluenitrile, 4-Tolyl cyanide, NSC 70985, p-Cyanotoluene, p-Methylbenzonitrile, p-Toluenenitrile, p-Tolyl cyanide, p-Tolylnitrile
InChI Formula InChI=1S/C8H7N/c1-7-2-4-8(6-9)5-3-7/h2-5H,1H3
SMILES Cc1ccc(cc1)C#N
IUPAC Name 4-methylbenzonitrile
Molecular Weight (g/mol) 117.15
Quantity 100 mg
Formula Weight 117.0578 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.