missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzyl Bromide, TRC

Product Code. 30471843
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471843 100 g 100g
30479639 5 g 5g
30474617 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30471843 Supplier Toronto Research Chemicals Supplier No. TRCM289735100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-81-4
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.06
Synonym alpha-Bromo-p-xylene, 1-(Bromomethyl)-4-methylbenzene, 4-(Bromomethyl)toluene, 4-Methylbenzyl Bromide, NSC 8050, p-(Bromomethyl)toluene, p-Methylbenzyl Bromide, p-Xylyl Bromide, p-Xylyl alpha-Bromide, alpha-Bromo-p-xylene, alpha-Bromo-p-xylol
IUPAC Name 1-(bromomethyl)-4-methylbenzene
SMILES Cc1ccc(CBr)cc1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C8H9Br/c1-7-2-4-8(6-9)5-3-7/h2-5H,6H2,1H3
Formula Weight 183.9888
Chemical Name or Material 4-Methylbenzyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.