missing translation for 'onlineSavingsMsg'
Learn More

4-Methylimidazole, TRC

Product Code. 30138654
Change view
Click to view available options
Quantity:
1 g
25 g
250 g
Unit Size:
1g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138654 250 g 250g
30008752 25 g 25g
17933425 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138654 Supplier Toronto Research Chemicals Supplier No. TRCM312990250G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 822-36-6
Molecular Formula C4 H6 N2
Molecular Weight (g/mol) 82.1
Synonym 1H-Imidazole, 4-methyl- (9CI), Imidazole, 4(or 5)-methyl- (6CI,7CI), Imidazole, 4-methyl- (8CI), 4(5)-Methylimidazole, 4(or 5)-Methylimidazole, 4-Methyl-1H-imidazole, 4-Methylimidazole, 5-Methyl-1H-imidazole, 5-Methylimidazole, NSC 40744
IUPAC Name 5-methyl-1H-imidazole
SMILES Cc1cnc[nH]1

Specifications

Quantity 250 g
InChI Formula InChI=1S/C4H6N2/c1-4-2-5-3-6-4/h2-3H,1H3,(H,5,6)
Formula Weight 82.0531
Chemical Name or Material 4-Methylimidazol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.