missing translation for 'onlineSavingsMsg'
Learn More

4-Methyloctane, TRC

Product Code. 30470066
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30470066 1 g 1g
30469919 500 mg 500mg
30478621 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30470066 Supplier Toronto Research Chemicals Supplier No. TRCM3306481G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2216-34-4
Molecular Formula C9H20
Molecular Weight (g/mol) 128.26
Synonym (±)-4-Methyloctane, NSC 23691
SMILES CCCCC(C)CCC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H20/c1-4-6-8-9(3)7-5-2/h9H,4-8H2,1-3H3
Chemical Name or Material 4-Methyloctane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.