missing translation for 'onlineSavingsMsg'
Learn More

4-Methylsulfonyl-2-nitrotoluene, 99%

Product Code. 11409437
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409437 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11409437 Supplier Thermo Scientific Alfa Aesar Supplier No. B21444.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1671-49-4 | C8H9NO4S | 215.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1671-49-4
Molecular Formula C8H9NO4S
Molecular Weight (g/mol) 215.22
MDL Number MFCD00047808
InChI Key OXBDLEXAVKAJFD-UHFFFAOYSA-N
Synonym 4-methylsulfonyl-2-nitrotoluene, 1-methyl-4-methylsulfonyl-2-nitrobenzene, 2-nitro-4-methylsulfonyltoluene, 4-mesyl-2-nitrotoluene, 4-methanesulfonyl-1-methyl-2-nitrobenzene, methyl 3-nitro-p-tolyl sulfone, benzene, 1-methyl-4-methylsulfonyl-2-nitro, 2-nitro-4-methylsulf, ksc532k5r, 4-methy-2-nitrosulfonyl toluene
PubChem CID 687125
SMILES CC1=CC=C(C=C1[N+]([O-])=O)S(C)(=O)=O

Specifications

Melting Point 119°C to 122°C
Quantity 10 g
IUPAC Name 1-methyl-4-methylsulfonyl-2-nitrobenzene
Formula Weight 215.22
Percent Purity 99%
Chemical Name or Material 4-Methylsulfonyl-2-nitrotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.