missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)phenol, 97%

Product Code. 11374856
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374856 1 g 1g
11364856 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374856 Supplier Thermo Scientific Alfa Aesar Supplier No. H32935.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 14763-60-1 | C7H8O3S | 172.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14763-60-1
Molecular Formula C7H8O3S
Molecular Weight (g/mol) 172.20
MDL Number MFCD00047806
InChI Key KECCFSZFXLAGJS-UHFFFAOYSA-N
Synonym 4-methylsulfonyl phenol, 4-methanesulfonylphenol, 4-methanesulfonyl-phenol, 4-hydroxyphenyl methyl sulfone, 4-methylsulphonylphenol, phenol, p-methylsulfonyl, phenol, 4-methylsulfonyl, p-hydroxyphenyl methyl sulfone, p-methylsulfonyl phenol, unii-f7bif7zqj9
PubChem CID 123350
IUPAC Name 4-methylsulfonylphenol
SMILES CS(=O)(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 90°C to 95°C
Quantity 1 g
Formula Weight 172.2
Percent Purity 97%
Chemical Name or Material 4-(Methylsulfonyl)phenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.