missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)benzenemethanol, TRC

Product Code. 30474816
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
missing translation for 'unitSize'
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474816 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30474816 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCM33128025G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Methylthio)benzenemethanol
CAS 3446-90-0
Molecular Formula C26 H33 F O8
Quantity 25 g
Synonym 4-(Hydroxymethyl)phenyl methyl Sulfide, 4-(Methylmercapto)benzyl alcohol, 4-(Methylthio)benzyl alcohol, 4-Methylsulfanylbenzyl alcohol, [4-(Methylsulfanyl)phenyl]methanol, p-(Methylthio)benzyl alcohol
InChI Formula InChI=1S/C26H33FO8/c1-13(28)33-12-20(32)26-21(34-22(2,3)35-26)10-15-16-9-18(30)17-8-14(29)6-7-23(17,4)25(16,27)19(31)11-24(15,26)5/h6-8,15-16,18-19,21,30-31H,9-12H2,1-5H3/t15-,16+,18-,19+,21+,23+,24+,25-,26+/m1/s1
SMILES CC(=O)OCC(=O)[C@]12OC(C)(C)O[C@H]1C[C@@H]3[C@@H]4C[C@@H](O)C5=CC(=O)C=C[C@]5(C)[C@]4(F)[C@@H](O)C[C@]23C
IUPAC Name [2-[(1S,2R,4S,8R,9S,11S,12S,13S,19R)-12-fluoro-11,19-dihydroxy-6,6,9,13-tetramethyl-16-oxo-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-dien-8-yl]-2-oxoethyl] acetate
Molecular Weight (g/mol) 492.534
Formula Weight 492.216
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.