Learn More
Melford 4-Methylumbelliferyl-2-acetamido-2-deoxy-6-sulfate-β-D-glucopyranoside Sodium Salt
Product Code. 30018501
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018501 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30018501 Supplier Melford Supplier No. M641500.025
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
Fluorescent substrate for N-Acetyl-β-D-glucosaminidase (Hexosaminidase A).
Fluorescent substrate for N-Acetyl-β-D-glucosaminidase (Hexosaminidase A). Allows more rapid detection of Tay-Sachs.
4-Methylumbelliferone < 50ppm
Nonsulfated Derivative: None Detected (NMR, TLC)
Checked by enzymatic assay
Specifications
Quantity 25 mg