missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl Caprylate, TRC

Product Code. 30134945
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134945 500 mg 500mg
30011280 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134945 Supplier Toronto Research Chemicals Supplier No. TRCM337645500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20671-66-3
Molecular Formula C18H22O4
Molecular Weight (g/mol) 302.36
Synonym 4-Methylumbelliferone Octanoate, 7-Hydroxy-4-methyl-coumarin Octanoate, 4-Methyl-2-oxo-2H-1-benzopyran-7-yl Ester Octanoic Acid
IUPAC Name 4-methyl-2-oxo-2H-chromen-7-yl octanoate
SMILES O=C1OC2=C(C(C)=C1)C=CC(OC(CCCCCCC)=O)=C2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C18H22O4/c1-3-4-5-6-7-8-17(19)21-14-9-10-15-13(2)11-18(20)22-16(15)12-14/h9-12H,3-8H2,1-2H3
Formula Weight 302.15
Chemical Name or Material 4-Methylumbelliferyl Caprylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.