Learn More
Melford 4-Methylumbelliferyl Elaidate
Product Code. 30018503
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 30018503
Brand: Melford M641650.1
Fluorescent substrate for Esterase.
Fluorescent substrate for Esterase. Purity: > 98%. 4-Methylumbelliferone < 50ppm. Soluble in 5% toluene.
Specifications
Quantity 100 mg