Learn More
Melford 4-Methylumbelliferyl Elaidate
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Fluorescent substrate for Esterase.
Fluorescent substrate for Esterase. Purity: > 98%. 4-Methylumbelliferone < 50ppm. Soluble in 5% toluene.
Specifications
Specifications
| Quantity | 100 mg |