Learn More
Melford 4-Methylumbelliferyl-α-L-fucopyranoside
Product Code. 30018507
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018507 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30018507 Supplier Melford Supplier No. M652000.025
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Fluorescent substrate for α-L-Fucosidase.
Fluorescent substrate for α-L-Fucosidase.
4-Methylumbelliferone <50ppm.
Specifications
Quantity 25 mg